Learn More
Invitrogen™ MAGEC3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MAGC3.
Specifications
Specifications
| Antigen | MAGEC3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MAGEC3 |
| Gene Accession No. | Q8TD91 |
| Gene Alias | cancer/testis antigen 7.2; cancer/testis antigen family 7, member 2; CT7.2; HCA2; hepatocellular carcinoma-associated antigen 2; hepatocellular carcinoma-associated protein HCA2; MAGE family member C3; MAGE family testis and tumor-specific protein; MAGEC3; MAGE-C3; MAGE-C3 antigen; MAGEC4; melanoma antigen family C, 3; melanoma antigen family C3; melanoma-associated antigen C3 |
| Gene Symbols | MAGEC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.