Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568045 Supplier Invitrogen™ Supplier No. PA568045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

MAGEC3 is a member of the MAGEC gene family. The members of this family are not expressed in normal tissues, except for testis, and are expressed in tumors of various histological types. The MAGEC genes are clustered on chromosome Xq26-q27. Two transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEC3
Gene Accession No. Q8TD91
Gene Alias cancer/testis antigen 7.2; cancer/testis antigen family 7, member 2; CT7.2; HCA2; hepatocellular carcinoma-associated antigen 2; hepatocellular carcinoma-associated protein HCA2; MAGE family member C3; MAGE family testis and tumor-specific protein; MAGEC3; MAGE-C3; MAGE-C3 antigen; MAGEC4; melanoma antigen family C, 3; melanoma antigen family C3; melanoma-associated antigen C3
Gene Symbols MAGEC3
Host Species Rabbit
Immunogen Synthesized peptide derived from human MAGEC3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139081
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.