Learn More
Invitrogen™ MAGED2 Polyclonal Antibody

Description
MAGED2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MAGED2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | MAGED2 |
| Gene Accession No. | Q9UNF1 |
| Gene Alias | 11B6; 4833439A22Rik; BARTS5; BCG1; BCG-1; Breast cancer-associated gene 1 protein; HCA10; hepatocellular carcinoma-associated protein JCL-1; JCL-1; MAGE family member D2; MAGED; MAGED2; MAGE-D2; MAGE-D2 antigen; melanoma antigen family D, 2; melanoma antigen family D2; melanoma antigen, family D, 2; melanoma-associated antigen D2; MGC8386; ORF1 |
| Gene Symbols | MAGED2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.