Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGED1 Monoclonal Antibody (353124)
GREENER_CHOICE

Catalog No. PIMA524096
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524096 Supplier Invitrogen™ Supplier No. MA524096
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 353124
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene MAGED1
Gene Accession No. Q9Y5V3
Gene Alias 11B6; 2810433C11Rik; 5430405L04Rik; BCG-1; Dlixin; Dlxin1; DLXIN-1; DXBwg1492e; JCL-1; MAGE family member D1; MAGE tumor antigen CCF; MAGED1; MAGE-D1; MAGE-D1 antigen; Maged2; melanoma antigen family D, 1; melanoma antigen family D1; melanoma antigen, family D, 1; melanoma-associated antigen D1; mFLJ00163; neurotrophin receptor-interacting MAGE homolog; NRAGE; PP2250; PRO2292; Sertoli cell necdin-related gene protein 1; SNERG-1
Gene Symbols MAGED1
Host Species Mouse
Immunogen E. coli-derived recombinant human NRAGE Met1-Glu778.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9500
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.