Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565365 Supplier Invitrogen™ Supplier No. PA565365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARPKSAFKVQN ATTKGPNGVY DFSQAHNAKD VPNTQPKAAF KSQNATPKGP NAAYDFSQAA TTGELAANKS EMAFKAQNAT TKVGPNATYN FSQSLNANDL ANSRPKTPFK AWNDTTKAPT ADTQTQNVNQ AKMATSQADI ETDPGISEP Highest antigen sequence identity to the following orthologs - mouse 67%, rat 68%.

This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGED1
Gene Accession No. Q9Y5V3
Gene Alias 11B6; 2810433C11Rik; 5430405L04Rik; BCG-1; Dlixin; Dlxin1; DLXIN-1; DXBwg1492e; JCL-1; MAGE family member D1; MAGE tumor antigen CCF; MAGED1; MAGE-D1; MAGE-D1 antigen; Maged2; melanoma antigen family D, 1; melanoma antigen family D1; melanoma antigen, family D, 1; melanoma-associated antigen D1; mFLJ00163; neurotrophin receptor-interacting MAGE homolog; NRAGE; PP2250; PRO2292; Sertoli cell necdin-related gene protein 1; SNERG-1
Gene Symbols MAGED1
Host Species Rabbit
Immunogen Recombinant Human MAGED1. Recombinant protein control fragment (Product #RP-104776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.