Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGED1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MAGED1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7247
![]() ![]() |
Proteintech Group Inc
115391AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MAGE-D2, also known as Melanoma-associated antigen D2, belongs to the melanoma antigen encoding gene (MAGE) family with a MAGE domain. Rat and mouse orthologues have been isolated. Compared to other proteins (MAGE-A, MAGE- B and MAGE-C clusters) MAGE-D2 is expressed ubiquitously. This intracellular protein shows high expression in specific brain regions and in the interstitium of testes. Though the exact function of this protein is yet to be deduced, MAGE-D2 is considered as a hot spot for X-linked mental retardation. Research reveals that MAGE-D2 might have some role in breast and gastric cancer, as well as in hepatocellular carcinoma. MAGE-D2 is also known to interact physically with p53 and modify its activity, thus acting as a p53-dissociator.Specifications
MAGED1 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9ES73, Q9QYH6, Q9Y5V3 | |
84469, 94275, 9500 | |
MAGED1 Fusion Protein Ag2109 | |
Primary | |
-20°C | |
MAGED1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DLXIN 1, MAGE D1 antigen, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, Melanoma associated antigen D1, NRAGE, PP2250 | |
Maged1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title