Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGED2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen MAGED2
Concentration 0.5 mg/mL
Dilution Western Blot 5-7 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224694SS
SDP
View Documents
Novus Biologicals
NBP224694SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224694
SDP
View Documents
Novus Biologicals
NBP224694
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

MAGED2 Polyclonal specifically detects MAGED2 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

MAGED2
Western Blot 5-7 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Canine, Equine, Primate
11B6MAGE-D2 antigen, BCG-1, BCG1MAGEDHCA10, breast cancer associated gene 1, Breast cancer-associated gene 1 protein, hepatocellular carcinoma associated protein, hepatocellular carcinoma-associated protein HCA10, Hepatocellular carcinoma-associated protein JCL-1, JCL-1, MAGE-D2, melanoma antigen family D, 2, melanoma-associated antigen D2, MGC8386
MAGED2
IgG
Protein A purified
This antibody may also recognize Maged1 as one of the Maged2 sequences used for immunogen is 94% homologous to Maged1.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cancer, Immunology
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
10916
human MAGE-D2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.