Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGED2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556782 Supplier Invitrogen™ Supplier No. PA556782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSDTSESGAG LTRFQAEASE KDSSSMMQTL LTVTQNVEVP ETPKASKALE VSEDVKVSKA SGVSKATEVS KTPEAREAPA TQASSTTQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.

This gene is a member of the MAGED gene family. The MAGED genes are clustered on chromosome Xp11. This gene is located in Xp11.2, a hot spot for X-linked intellectual disability (XLID). Mutations in this gene cause a form of transient antenatal Bartter's syndrome. This gene may also be involved in several types of cancer, including breast cancer and melanoma. The protein encoded by this gene is progressively recruited from the cytoplasm to the nucleoplasm during the interphase and after nucleolar stress and is thus thought to play a role in cell cycle regulation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGED2
Gene Accession No. Q9UNF1
Gene Alias 11B6; 4833439A22Rik; BARTS5; BCG1; BCG-1; Breast cancer-associated gene 1 protein; HCA10; hepatocellular carcinoma-associated protein JCL-1; JCL-1; MAGE family member D2; MAGED; MAGED2; MAGE-D2; MAGE-D2 antigen; melanoma antigen family D, 2; melanoma antigen family D2; melanoma antigen, family D, 2; melanoma-associated antigen D2; MGC8386; ORF1
Gene Symbols MAGED2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGED2. Recombinant protein control fragment (Product #RP-95270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.