Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGED2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571736
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1736 10 μL
50-157-1735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1736 Supplier Bethyl Laboratories, Inc Supplier No. IHC00237T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

MAGE-D2, also known as Melanoma-associated antigen D2, belongs to the melanoma antigen encoding gene (MAGE) family with a MAGE domain. Rat and mouse orthologues have been isolated. Compared to other proteins (MAGE-A, MAGE- B and MAGE-C clusters) MAGE-D2 is expressed ubiquitously. This intracellular protein shows high expression in specific brain regions and in the interstitium of testes. Though the exact function of this protein is yet to be deduced, MAGE-D2 is considered as a hot spot for X-linked mental retardation. Research reveals that MAGE-D2 might have some role in breast and gastric cancer, as well as in hepatocellular carcinoma. MAGE-D2 is also known to interact physically with p53 and modify its activity, thus acting as a p53-dissociator.

Specifications

Antigen MAGED2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAGED2
Gene Accession No. Q9UNF1
Gene Alias 11B6, BARTS5, BCG1, BCG-1, breast cancer associated gene 1, breast cancer-associated gene 1 protein, HCA10, hepatocellular carcinoma associated protein, hepatocellular carcinoma-associated protein HCA10, hepatocellular carcinoma-associated protein JCL-1, JCL-1, MAGED, MAGE-D2, MAGE-D2 antigen, melanoma antigen family D, 2, melanoma antigen family D2, melanoma-associated antigen D2, MGC8386
Gene Symbols MAGED2
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10916
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.