Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100735 Supplier Invitrogen™ Supplier No. PA5100735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAGD4.

May enhance ubiquitin ligase activity of RING-type zinc finger-containing E3 ubiquitin-protein ligases. Proposed to act through recruitment and/or stabilization of the Ubl-conjugating enzyme (E2) at the E3:substrate complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGED4
Gene Accession No. Q96JG8
Gene Alias KIAA1859; MAGE family member D4; MAGE1; MAGED4; MAGE-D4 antigen; MAGED4A; MAGED4B; MAGEE1; MAGE-E1; MAGE-E1 antigen; melanoma antigen family D, 4; melanoma antigen family D4; melanoma-associated antigen D4
Gene Symbols MAGED4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGED4; MAGED4B(Accession Q96JG8), corresponding to amino acid residues G577-H627.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728239
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.