Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551961 Supplier Invitrogen™ Supplier No. PA551961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEGSFSVQSE SYSVEDMDEG SDEVGEEEMV EGNDYEEFGA FGGYGTLTSF DIHILRAFGS LGPGLRILSN EPWELENPVL AQTLVEALQL DPETLANETA ARAANVARAA ASNRAA Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 26%.

May enhance ubiquitin ligase activity of RING-type zinc finger-containing E3 ubiquitin-protein ligases. Proposed to act through recruitment and/or stabilization of the Ubl-conjugating enzyme (E2) at the E3:substrate complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGED4
Gene Accession No. Q96JG8
Gene Alias KIAA1859; MAGE family member D4; MAGE1; MAGED4; MAGE-D4 antigen; MAGED4A; MAGED4B; MAGEE1; MAGE-E1; MAGE-E1 antigen; melanoma antigen family D, 4; melanoma antigen family D4; melanoma-associated antigen D4
Gene Symbols MAGED4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGED4. Recombinant protein control fragment (Product #RP-102110).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728239
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.