Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGED4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548581
Description
May enhance ubiquitin ligase activity of RING-type zinc finger-containing E3 ubiquitin-protein ligases. Proposed to act through recruitment and/or stabilization of the Ubl-conjugating enzyme (E2) at the E3:substrate complex.
Specifications
MAGED4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96JG8 | |
MAGED4 | |
KLH conjugated synthetic peptide between 328-357 amino acids from the Central region of human MAGED4B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MAGED4 | |
Melanoma-associated antigen D4, MAGE-D4 antigen, MAGE-E1 antigen, MAGED4, KIAA1859, MAGED4A, MAGEE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728239 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction