Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103463 Supplier Invitrogen™ Supplier No. PA5103463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAGE1.

This gene encodes an alpha-dystrobrevin-associated MAGE (melanoma-associated antigen) protein, which is a member of the MAGE family. The protein contains a nuclear localization signal in the N-terminus, 30 12-amino acid repeats beginning at nt 60 with the consensus sequence ASEGPSTSVLPT, and two MAGE domains in the C-terminus. It may play a signaling role in brain, muscle, and peripheral nerve. This gene is located on X chromosome in a region containing loci linked to cognitive disability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEE1
Gene Accession No. A1A5P9, Q6PCZ4, Q9HCI5
Gene Alias AI847422; Alpha-dystrobrevin-associated MAGE Protein; DAMAGE; dystrobrevin-associated MAGE protein; HCA1; hepatocellular carcinoma-associated HCA1; hepatocellular carcinoma-associated protein 1; hepatocellular carcinoma-associated protein HCA1; KIAA1587; MAGE family member E1; Magee1; MAGE-E1 antigen; melanoma antigen family E, 1; melanoma antigen family E1; melanoma antigen, family E, 1; melanoma-associated antigen E1; mMage-e1; RGD1560259
Gene Symbols MAGEE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGEE1(Accession Q9HCI5), corresponding to amino acid residues E716-I766.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107528, 317232, 57692
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.