Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102024 Supplier Invitrogen™ Supplier No. PA5102024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAGEC2.

MAGE-C2/C3 (Melanoma-associated antigen C2, Hepatocellular carcinoma-associated antigen 587) is a 373 amino acid protein encoded by the human gene MAGEC2. Genes of the MAGE family direct the expression of tumor antigens that are recognized on human melanomas by autologous cytolytic T lymphocytes. Among tumor tissues, MAGE-C2/C3 is frequently expressed in seminomas, melanomas, and bladder transitional cell carcinomas. It is also expressed in a significant fraction of head and neck carcinomas, breast carcinomas, nonsmall-cell lung carcinomas, and sarcomas. In hepatocellular carcinoma, there is a inverse correlation between tumor differentiation and protein expression, i. e. the lower the differentiation, the higher percentage of expression. MAGE-C2/C3 is not expressed in normal tissues, except in germ cells in the seminiferous tubules and in Purkinje cells of the cerebellum. It is strongly expressed in spermatogonia and primary spermatocytes, however, at later stages of maturation, expression gradually decreases to become undetectable in mature spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEC2
Gene Accession No. Q9UBF1
Gene Alias Cancer/testis antigen 10; CT10; HCA587; hepatocellular cancer antigen 587; hepatocellular carcinoma-associated antigen 587; MAGE family member C2; MAGEC2; MAGE-C2; MAGE-C2 antigen; MAGEE1; MAGE-E1 antigen; melanoma antigen family C, 2; melanoma antigen family C2; melanoma antigen, family E, 1, cancer/testis specific; melanoma-associated antigen C2; uncharacterized protein LOC317590
Gene Symbols MAGEC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGEC2(Accession Q9UBF1), corresponding to amino acid residues G194-A244.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51438
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.