Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556525 Supplier Invitrogen™ Supplier No. PA556525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALKFVSDAH DEEPWSWPEE YNKALEGDKT KERSLTAGLE FWSEDTMNDK ANDLVQLAIS VTEEMLPIHQ DELLAHTGKE FEDVFPNILN RATLILDMFY GLSLIEVDT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes a member of the E subfamily of MAGE (melanoma antigen-encoding gene) gene family. The gene is intronless and the encoded protein has two of the MAGE domains which are characteristic of MAGE family proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEE2
Gene Accession No. Q8TD90
Gene Alias 9630059J11Rik; BB131182; HCA3; Hepatocellular carcinoma-associated protein 3; hepatocellular carcinoma-associated protein HCA3; MAGE family member E2; MAGEE2; Mage-e2; MAGE-E2 antigen; melanoma antigen family E, 2; melanoma antigen family E2; melanoma antigen, family E, 2; melanoma-associated antigen E2; RGD1564872
Gene Symbols MAGEE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEE2. Recombinant protein control fragment (Product #RP-91970).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139599
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.