Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEE2 Polyclonal Antibody

Catalog No. PIPA535919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535919 Supplier Invitrogen™ Supplier No. PA535919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (89%), Rat (87%), Bovine (92%).

This gene encodes a member of the E subfamily of MAGE (melanoma antigen-encoding gene) gene family. The gene is intronless and the encoded protein has two of the MAGE domains which are characteristic of MAGE family proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEE2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene MAGEE2
Gene Accession No. Q8TD90
Gene Alias 9630059J11Rik; BB131182; HCA3; Hepatocellular carcinoma-associated protein 3; hepatocellular carcinoma-associated protein HCA3; MAGE family member E2; MAGEE2; Mage-e2; MAGE-E2 antigen; melanoma antigen family E, 2; melanoma antigen family E2; melanoma antigen, family E, 2; melanoma-associated antigen E2; RGD1564872
Gene Symbols MAGEE2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human MAGEE2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139599, 302392
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.