Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513166
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513166 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513166 Supplier Invitrogen™ Supplier No. PA513166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGEH1 is not fully elucidated, it may play a role in embryonal development and tumor transformation or aspects of tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAGEH1
Gene Accession No. Q9H213
Gene Alias 1-Apr; 2010107K23Rik; Apoptosis-related protein 1; APR1; APR-1; MAGE family member H1; MAGEH; Mageh1; MAGE-H1 antigen; melanoma antigen family H, 1; melanoma antigen family H1; melanoma antigen, family H, 1; melanoma-associated antigen H1; Restin
Gene Symbols MAGEH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 186-218 amino acids from the C-terminal region of human MAGEH1.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.