Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570460 Supplier Invitrogen™ Supplier No. PA570460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Dog: 100%; Human: 100%; Pig: 88%; Rabbit: 100%.

MAGEL2 gene ontology annotations related to this gene include negative regulation of transcription, DNA-templated; positive regulation of actin nucleation; regulation of circadian rhythm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAGEL2
Gene Accession No. Q9UJ55
Gene Alias LOW QUALITY PROTEIN: MAGE-like protein 2; MAGE family member L2; MAGEL2; Mage-l2; MAGE-like; MAGE-like 2; MAGE-like protein 2; melanoma antigen family L 2; melanoma antigen family L2; melanoma antigen, family L, 2; melanoma antigen-like gene 2; melanoma antigen-like protein 2; NDNL1; necdin-like 1; necdin-like protein 1; nM15; ns7; Protein nM15; protein nS7; PWLS; SHFYNG
Gene Symbols MAGEL2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human MAGEL2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54551
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.