Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA125136
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA125136 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA125136 Supplier Invitrogen™ Supplier No. PA125136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: rat brain. PA1-25136 is expected to cross react with human and mouse due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Cell culture media with no preservative
Gene Magi1
Gene Accession No. Q4L1J4, Q96QZ7
Gene Alias AIP3; AIP-3; Atrophin-1-interacting protein 3; BAI1-associated protein 1; Baiap1; BAP1; BAP-1; Gukmi1; MAGI1; Magi-1; MAGI1c; Magi1d; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase inverted 1; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 1; mKIAA4129; TNRC19; Trinucleotide repeat-containing gene 19 protein; WW domain-containing protein 3; WWP3
Gene Symbols Magi1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 282-296 of MAGI-1 with a carboxy terminal added cysteine residue conjugated to maleimide-activated KLH.
Purification Method IgG fraction
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500261, 9223
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.