Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527743
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (96%), Rat (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
MAGI1 | |
Polyclonal | |
Unconjugated | |
Magi1 | |
AIP3; AIP-3; Atrophin-1-interacting protein 3; BAI1-associated protein 1; Baiap1; BAP1; BAP-1; Gukmi1; MAGI1; Magi-1; MAGI1c; Magi1d; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase inverted 1; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 1; mKIAA4129; TNRC19; Trinucleotide repeat-containing gene 19 protein; WW domain-containing protein 3; WWP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14924, 9223 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
Q6RHR9, Q96QZ7 | |
Magi1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human MAGI1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction