Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599245 Supplier Invitrogen™ Supplier No. PA599245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAGI2.

Seems to act as scaffold molecule at synaptic junctions by assembling neurotransmitter receptors and cell adhesion proteins. May play a role in regulating activin-mediated signaling in neuronal cells. Enhances the ability of PTEN to suppress AKT1 activation. Plays a role in nerve growth factor (NGF)-induced recruitment of RAPGEF2 to late endosomes and neurite outgrowth. Florindo C.S., Submitted (JUL-2003) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004). Devroe E., J. Biol. Chem. 279:24444-24451(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGI2
Gene Accession No. O88382, Q86UL8, Q9WVQ1
Gene Alias activin receptor interacting protein 1; activin receptor-interacting protein 1; Acvri1; ACVRINP1; Acvrip1; AIP1; AIP-1; ARIP1; Atrophin-1-interacting protein 1; atrophin-1-interacting protein A; KIAA0705; MAGI2; MAGI-2; membrane associated guanylate kinase, WW and PDZ domain containing 2; membrane-associated guanylate kinase inverted 2; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2; mKIAA0705; Sscam; S-SCAM; Synaptic-scaffolding molecule
Gene Symbols MAGI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGI2(Accession Q86UL8), corresponding to amino acid residues E235-E285.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113970, 50791, 9863
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.