Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531449 Supplier Invitrogen™ Supplier No. PA531449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, HeLa. Predicted reactivity: Mouse (96%), Rat (95%), Chicken (93%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Membrane-associated guanylate kinase with inverted orientation (MAGI3) is a novel, 130 kDa guanylate kinase which is part of a family of multi-PDZ domain containing guanylate kinases. These guanylate kinases are localized to epithelial cell tight junctions. MAGI3 and PTEN/MMAC cooperate to modulate the kinase activity of AKT/protein kinase B (PKB). MAGI3 positions the PTEN/MMAC phosphatase to specific subcellular locations that are involved with the regulation of cell proliferation and survival. MAGI3, as well as other proteins in this family, are made up of an N-terminal guanylate kinase domain, followed by a WW domain, named for two conserved tryptophan residues and first identified as two repeats in mouse YAP65, which is them followed by 5 PDZ domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI3
Applications Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol; pH 7
Gene MAGI3
Gene Accession No. Q5TCQ9
Gene Alias 4732496O19Rik; 6530407C02Rik; AA407180; AI120132; dJ730K3.2; GKWW domain; KIAA1634; Magi3; MAGI-3; membrane associated guanylate kinase, WW and PDZ domain containing 3; membrane-associated guanylate kinase inverted 3; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 3; membrane-associated guanylate kinase-related (MAGI-3); membrane-associated guanylate kinase-related 3; mKIAA1634; PDZ 4-5; RP4-730K3.1; scaffolding protein SLIPR; scaffolding-like protein; Slipr
Gene Symbols MAGI3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MAGI3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.