Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGI3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS18629R

Description
Membrane-associated guanylate kinase with inverted orientation (MAGI3) is a novel, 130 kDa guanylate kinase which is part of a family of multi-PDZ domain containing guanylate kinases. These guanylate kinases are localized to epithelial cell tight junctions. MAGI3 and PTEN/MMAC cooperate to modulate the kinase activity of AKT/protein kinase B (PKB). MAGI3 positions the PTEN/MMAC phosphatase to specific subcellular locations that are involved with the regulation of cell proliferation and survival. MAGI3, as well as other proteins in this family, are made up of an N-terminal guanylate kinase domain, followed by a WW domain, named for two conserved tryptophan residues and first identified as two repeats in mouse YAP65, which is them followed by 5 PDZ domains.Specifications
MAGI3 | |
Polyclonal | |
Unconjugated | |
MAGI3 | |
dJ730K3.2; GKWW domain; KIAA1634; Magi3; MAGI-3; membrane associated guanylate kinase, WW and PDZ domain containing 3; Membrane-associated guanylate kinase inverted 3; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 3; membrane-associated guanylate kinase-related (MAGI-3); membrane-associated guanylate kinase-related 3; PDZ 4-5; RP4-730K3.1; scaffolding protein SLIPR; Scaffolding-like protein; Slipr | |
Rabbit | |
Protein A | |
RUO | |
260425 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q5TCQ9 | |
MAGI3 | |
KLH conjugated synthetic peptide derived from human MAGI3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction