Learn More
Invitrogen™ MagicMark™ XP Western Protein Standard
Description
Includes
50 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue
- MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
- The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
- The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
- The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
- Store at -20°C
Applications
- Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
- Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | 50 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue. Store at -20°C. |
| Detection Method | User defined detection system |
| Number of Markers | 9 |
| Product Type | Protein Ladder |
| Ready to Load | Yes |
| Size Range | 20 to 220 kDa |
| Stain Type | Unstained |
| Gel Compatibility | Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels |
| Molecular Weight (g/mol) | 220, 120, 100, 80, 60, 50, 40, 30, 20 kDa |
| Product Line | MagicMark |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.