Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magmas Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Magmas |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Magmas Polyclonal specifically detects Magmas in Human, Mouse samples. It is validated for Western Blot.Specifications
Magmas | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
51025 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
MAGMAS, mitochondria associated protein involved in granulocyte macrophage colonystimulating factor signal transduction, Mitochondria-associated granulocyte macrophage CSF-signaling molecule, mitochondrial import inner membrane translocase subunit TIM16, presequence translocase-associated motor 16 homolog (S. cerevisiae), Presequence translocated-associated motor subunit PAM16, TIM16, TIMM16magmas-like protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-125 of human Magmas (NP_057153.8). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title