Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magmas Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260718
Description
Magmas Polyclonal specifically detects Magmas in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Magmas | |
Polyclonal | |
Immunoprecipitation 8-15 ul/mg lysate | |
MAGMAS, mitochondria associated protein involved in granulocyte macrophage colonystimulating factor signal transduction, Mitochondria-associated granulocyte macrophage CSF-signaling molecule, mitochondrial import inner membrane translocase subunit TIM16, presequence translocase-associated motor 16 homolog (S. cerevisiae), Presequence translocated-associated motor subunit PAM16, TIM16, TIMM16magmas-like protein | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
PAM16 | |
between 50 and 100 | |
100 μL | |
51025 | |
Store at 4degreesC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction