Learn More
Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit
Description
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.
Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• Compatible with multiple downstream assays
• Single purification protocol for routinely tested pathogens and respiratory biological specimen without need for additional lysis
• Ability to purify various samples on single plate
• Sustainable product with biodegradable ingredients
• Designed to work with KingFisher Apex Dx instrument in under 40 mins
The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic-bead purification and in vitro diagnostic procedures.
Specifications
Specifications
| Content And Storage | • Binding Solution, 160 mL |
| Format | 96-well Plate |
| Isolation Technology | Magnetic Bead |
| Sample Type | Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum |
| For Use With (Equipment) | KingFisher™ Apex Dx |
| For Use With (Application) | Real-Time PCR |
| No. of Reactions | Up to 600 preps |
| Nucleic Acid Type | Viral and Bacterial |
| Product Line | MagMAX™ |
| Quantity | Up to 600 preps |
| Show More |
For In Vitro Diagnostic Use
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.