Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosytstems™ MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer

Catalog No. p-200007342
Encompass_Preferred
Click to view available options
No. of Reactions:
Up to 100 Reactions
Up to 1000 Reactions

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi.

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic
Color Clear
Content And Storage Store at –15 to –25°C.
Format Bottle
Isolation Technology Beads
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions Up to 100 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX™
Product Type Reagent
Quantity 5 mL
Sample Volume 200–400 μL
Shipping Condition Dry Ice
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.