Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MagnaBind™ Carboxyl Derivatized Beads

Catalog No. PI21353 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21353 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21353 Supplier Thermo Scientific™ Supplier No. 21353
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.

Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

MagnaBind Carboxyl Derivatized Beads are supplied as an aqueous suspension of magnetic iron oxide beads coated with carboxyl groups for covalent coupling of molecules using EDC, a zero-length crosslinker that is amine- and carboxyl-reactive. Additionally, carboxyl derivatized beads have been used to purify plasmid DNA from cells.

Features of MagnaBind Carboxyl Derivatized Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: ∼20 mg/mL

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods
TRUSTED_SUSTAINABILITY

Specifications

Concentration Slurry: ∼20 mg/mL
Content And Storage Upon receipt store product at 4°C. Do not freeze product. Product is shipped at ambient temperature.
Description MagnaBind Carboxyl Derivatized Beads
Color Brown
For Use With (Application) Affinity Purification
Product Type Carboxyl Derivatized Bead
Product Line MagnaBind
Quantity 5 mL
Surface Functionality Carboxylic Acid
Volume (Metric) 5 mL
Diameter (Metric) 1 to 4 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.