Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Plate Clamp 96

Catalog No. PRV8251 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Plate Clamp 96
Plate Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRV8251 Plate Clamp 96
PRV8261 Plate Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8251 Supplier Promega Supplier No. V8251
Add to Cart
Add to Cart

Recommended for automated protocols. Designed to ensure that nonskirted PCR plates are uniformly flat for liquid transfer on a robotic platform.

  • Designed to ensure that nonskirted PCR plates are uniformly flat for liquid transfer on a robotic platform
  • Recommended for automated protocols

†Patent Pending.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Non-skirted plates and strip tubes
Product Type Plate Clamp 96
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.