Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium aluminum oxide, 99% (metals basis)

Catalog No. p-7049800
Click to view available options
Quantity:
100 g
500 g

CAS: 12068-51-8 | Al2MgO4 | 142.26 g/mol

Magnesium aluminum oxide is used in chemical research, chemicals, pharmaceutical research, pharmaceutical intermediates reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-51-8
Molecular Formula Al2MgO4
Molecular Weight (g/mol) 142.26
MDL Number MFCD00049475
InChI Key UAMZXLIURMNTHD-UHFFFAOYSA-N
Synonym alumina magnesia, cernel i, dialuminum magnesium tetraoxide, magnesium aluminate mgal2o4, dialuminum;magnesium;oxygen 2-, magnesium aluminate mg alo2 2, aluminum magnesium oxide, aluminum magnesium oxide mgal2o4, magnesium aluminum oxide mgal2o4, aluminate alo21-, magnesium 2:1

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Not soluble in water.
Formula Weight 142.26
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Magnesium aluminum oxide
EINECSNumber 235-100-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.