Alfa Aesar™ Magnesium bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 46667MD

Quantity 250mg

 View more versions of this product

Catalog No. AA46667MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 133395-16-1
    Molecular Formula C4F12MgN2O8S4
    Linear Formula [(CF3SO2)2N]2Mg
    Formula Weight 584.58
    MDL Number MFCD01321278
    UN Number UN1759
    Synonym Magnesium triflimide; Magnesium bis(trifluoromethanesulfonimide)
    Chemical Name or Material Magnesium bis (trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 584.58
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    CAS 133395-16-1
    UN Number UN1759
    Sensitivity Moisture Sensitive
    Molecular Formula C4F12MgN2O8S4
    Linear Formula [(CF3SO2)2N]2Mg
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point >200°C
    Quantity 250mg
    MDL Number MFCD01321278
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Color White to pale yellow
    Synonym Magnesium triflimide; Magnesium bis(trifluoromethanesulfonimide)
    Chemical Name or Material Magnesium bis (trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit