Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium boride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8814922

Quantity 100g

 View more versions of this product

Catalog No. AA8814922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12007-25-9
MgB2
MFCD00049752
Magnesium boride
B2Mg
45.93
Magnesium diboride

Specifications

12007-25-9
B2Mg
45.93
Magnesium diboride
Odorless
800°C (dec.)
Magnesium boride
Powder
MgB2
MFCD00049752
-100 Mesh
Dark Gray
100g
99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302;
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OM3078000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit