Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%

Catalog No. p-4781980
Click to view available options
Quantity:
100 g
500 g

CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

Magnesium carbonate hydroxide tetrahydrate is used as a mixture with huntite and as filler in polymers. It acts as a flame retardant and fire retardant additive for polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
Synonym Magnesium carbonate, basic, Magnesium hydroxide carbonate
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range MgO approx.40-43.5%
Solubility Information Slightly soluble in water.
Formula Weight 467.67 (395.58 Anhydrous)
Grade Reagent
Odor Odorless
Chemical Name or Material Magnesium carbonate hydroxide tetrahydrate
EINECSNumber 235-192-7
RTECSNumber FG1797000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.