Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, 1 M Solution, Sterile-Filtered, PROMO, Thermo Scientific Chemicals

Catalog No. J61014AKPM
Click to view available options
Quantity:
250 mL

Applications

Magnesium chloride, 1M aq. solution, sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

Solubility

Soluble in water (542 mg/mL at 20°C), alcohol, methanol (15.5 g/100 g at 0°C), and ethanol (3.61 g/100 g at 0°C).

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Very hygroscopic.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21 g/mol
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
IUPAC Name magnesium(2+) dichloride
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water, alcohol, methanol, and ethanol.
Concentration or Composition (by Analyte or Components) 1 M
Chemical Name or Material Magnesium chloride

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.