Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride, anhydrous, 99%

Catalog No. p-4407599
Click to view available options
Quantity:
1 kg
5 kg
25 kg

For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Hygroscopic; May contain trace amount of Zr as black insoluble particles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Melting Point 714°C
Color White to Yellow
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99%
Solubility Information Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine.
IUPAC Name magnesium(2+) dichloride
Formula Weight 95.22
Odor Odorless
Sensitivity Hygroscopic
Appearance White to yellow crystals
Density 2.32 g/cm3 at 20°C
Chemical Name or Material Magnesium Chloride, Anhydrous
Show More Show Less
EINECSNumber 232-094-6
RTECSNumber OM2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.