Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium chloride, anhydrous, 99%
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Hygroscopic; May contain trace amount of Zr as black insoluble particles

Specifications
Specifications
Melting Point | 714°C |
Color | White to Yellow |
Physical Form | Crystals |
Quantity | 5 kg |
Assay Percent Range | 99% |
Solubility Information | Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine. |
IUPAC Name | magnesium(2+) dichloride |
Formula Weight | 95.22 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
EINECSNumber | 232-094-6 |
RTECSNumber | OM2800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction