Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride hexahydrate, 97+%

Catalog No. p-4407598
Click to view available options
Quantity:
500 g
2 kg
10 kg

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

It is utilized in the polymerase chain reactions (PCR). It is used for dust control, ice control, and road stabilization. It is employed in a variety of applications such as fertilizers, mineral supplements for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Pharmaceutical preparations are used in several medicines and topical or skin related applications. It acts as a coagulant in the preparation of tofu from soy milk.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C (decomposition)
Color White
Physical Form Flakes
Quantity 500 g
Assay Percent Range >98%
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White flakes
Density 1.57 g/cm3 at 20°C
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less
EINECSNumber 232-094-6
RTECSNumber OM2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.