Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride hexahydrate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
7791-18-6
Cl2Mg·6H2O
203.31
MFCD00149781
7791-18-6
Cl2Mg·6H2O
203.31
MFCD00149781
Aluminum (Al) 1ppm max.
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula MgCl2·6H2O
Ammonium 50ppm max.
Fieser 10,251; 15,196
Merck Index 15, 5726
Arsenic (As) 2ppm max.
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Barium (Ba) 20ppm max.
Formula Weight 203.31
Physical Form Crystalline Powder
Percent Purity 99%
Grade Analysis
Calcium (Ca) 1000ppm max.
Copper (Cu) 10ppm max.
Free Alkali 0.01% max. (MgO)
Free Acid 0.02% max.
Iron (Fe) 5ppm max.
Lead (Pb) 10ppm max.
Packaging Plastic drum
Potassium (K) 3000ppm max.
Sodium (Na) 5000ppm max.
Solubility Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Sulfate 100ppm max.
Trace Analysis 10ppm max.
Color White
Melting Point 117.0°C
Quantity 5kg
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning