Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate Technical, Reagents

Catalog No. 01337192
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.

Catalog No. 01-337-192

Supplier: Reagents Holdings Llc C223850010KG

Only null left
Add to Cart
Add to Cart

Magnesium Chloride Hexahydrate Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium dichloride hexahydrate
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade Technical
Packaging Poly Pail
Specific Gravity 1.56
Chemical Name or Material Magnesium Chloride Hexahydrate Technical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.