Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium ethoxide, Mg 21-22%

Catalog No. p-7049411
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1426430 250 g
AA14264A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1426430 Supplier Thermo Scientific Chemicals Supplier No. 01426430
Only null left
Add to Cart

CAS: 2414-98-4 | C4H10MgO2 | 114.427 g/mol

Magnesium ethoxide is used as olefin polymerization catalyst carrier for polypropylene, polyethylene and precision ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2414-98-4
Molecular Formula C4H10MgO2
Molecular Weight (g/mol) 114.427
MDL Number MFCD00009060
InChI Key XDKQUSKHRIUJEO-UHFFFAOYSA-N
Synonym magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
PubChem CID 164963
IUPAC Name magnesium;ethanolate
SMILES CC[O-].CC[O-].[Mg+2]

Specifications

Melting Point 270°C (decomposition)
Color White
Density 1.01 g/mL
Odor Odorless
Linear Formula Mg(OCH2CH3)2
Quantity 250 g
UN Number UN3205
Beilstein 3678841
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 114.44
Concentration or Composition (by Analyte or Components) Mg 21 to 22%
Physical Form Granules
Chemical Name or Material Magnesium ethoxide
Show More Show Less
Hazard Category H228-H319-H500-X
Hazard Statement GHS H Statement
H251-H319
Self-heating: may catch fire.
Causes serious eye irritation.
Precautionary Statement P210-P235+P410-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
DOTInformation Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
EINECSNumber 219-323-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.