Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hydroxide, 95-100.5%

Catalog No. AA1236736
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes | CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

It is employed as an additive in certain refractories. It is applied as a non-hazardous alkali in neutralizing acidic wastewater. It is also used in making artificial reefs. It widely used in milk of magnesia antacids and laxatives. Also works as an antiperspirant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 360°dec
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 95-100.5%
Solubility Information Soluble in dilute acids. Slightly soluble in water.
IUPAC Name magnesium(2+) dihydroxide
Formula Weight 58.33
Odor Odorless
Sensitivity Air Sensitive
Appearance White powder
Density 2.36 g/cm3 at 20°C
Chemical Name or Material Magnesium hydroxide
Show More Show Less
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber 215-170-3
RTECSNumber OM3570000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.