Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium methoxide, 7-8% in methanol

Catalog No. AA40448A1
Click to view available options
Quantity:
1 kg
2 kg

MgO films | CAS: 109-88-6

The use of magnesium methoxide for the deprotection of alkyl esters is described. This mild reagent provides a good method to cleave esters efficiently and more importantly, allows for effective differentiation between two different esters. Dimethyl carbonate is synthesized directly from carbon dioxide and methanol in the presence of magnesium methoxide. 20 mol% DEA additions to magnesium methoxide showed enhanced stability such that a time-dependent change in the sol was not observed in air. Also enhanced the crystallization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-88-6
MDL Number MFCD00053671

Specifications

Color Colorless to Yellow
Density 0.815 g/mL
Flash Point 14°C (57°F)
Odor Alcohol-like
Refractive Index 1.338
Quantity 1 kg
UN Number UN1230
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 86.38
Concentration or Composition (by Analyte or Components) 7 to 8% in methanol
Physical Form Liquid
Chemical Name or Material Magnesium methoxide
Show More Show Less
Hazard Category H225-H301+H311+H331-H370-X
Hazard Statement GHS H Statement
H225-H311-H331-H370-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3
EINECSNumber 203-715-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.