Learn More
Magnesium nitrate hexahydrate, 99.97% (metals basis), Thermo Scientific Chemicals
Description
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 330°C (decomposition) |
Melting Point | 89°C |
Physical Form | Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.97% (metals basis) |
Solubility Information | Very soluble in water,alcohol and liquid ammonia. |
Formula Weight | 256.41 (148.32 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 1.46 g/mL |
Chemical Name or Material | Magnesium nitrate hexahydrate |
Safety and Handling
EINECSNumber | 233-826-7 |
RTECSNumber | OM3756000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.