Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium perchlorate hexahydrate, 99% (metals basis)

Catalog No. p-7048839
Click to view available options
Quantity:
250 g
1 kg

CAS: 13446-19-0 | Cl2H12MgO14 | 331.287 g/mol

Commercially available magnesium perchlorate is reported as an extremely efficient catalyst for the synthesis of α-aminophosphonates. Magnesium perchlorate has been found to be an efficient catalyst for the synthesis of imines and phenylhydrazones by the reaction of carbonyl compounds with amines and phenylhydrazine in high yields at room temperatures and in short times.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-19-0
Molecular Formula Cl2H12MgO14
Molecular Weight (g/mol) 331.287
MDL Number MFCD00149778
InChI Key MGBDNCBRZIMSDZ-UHFFFAOYSA-L
Synonym magnesium perchlorate hexahydrate, acmc-20akl9, 2clo4.mg.6h2o, ksc491a3f, magnesium 2+ ion hexahydrate diperchlorate, magnesium 2+ hexahydrate diperchlorate
PubChem CID 16211969
IUPAC Name magnesium;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 185°C to 190°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99% (Metals basis)
UN Number UN1475
Merck Index 14,5678
Solubility Information Insoluble in water.
Formula Weight 331.33 (223.23 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Magnesium perchlorate hexahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: MAGNESIUM PERCHLORATE
EINECSNumber 233-108-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.