Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium phosphate, tribasic hydrate, 98+%, pure

Catalog No. AC391940010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391940010 1 kg Plastic bottle
AC391942500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391940010 Supplier Thermo Scientific Chemicals Supplier No. 391940010
Add to Cart

CAS: 53408-95-0 | Mg3O8P2 | 262.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53408-95-0
Molecular Formula Mg3O8P2
Molecular Weight (g/mol) 262.86
MDL Number MFCD00150251
InChI Key GVALZJMUIHGIMD-UHFFFAOYSA-H
Synonym magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
PubChem CID 6096954
SMILES [Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98+%
Solubility Information Solubility in water: insoluble.
IUPAC Name trimagnesium;diphosphate;hydrate
Formula Weight 262.86
Percent Purity ≥98%
Grade Pure
Loss on Ignition 32.0% max. (425°C)
Packaging Plastic bottle
Chemical Name or Material Magnesium phosphate, tribasic hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.