Learn More
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Description
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Quantity | 4 x 500 g |
Mesh Size | -325 Mesh |
Form | Powder |
Assay Percent Range | 99.8% |
Formula Weight | 24.3 |
Chemical Name or Material | Magnesium |
Safety and Handling
Hazard Category | H250-H260-H500 |
Precautionary Statement | P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c |
DOTInformation | Hazard Class: 4.3; Packaging Group: II |
EINECSNumber | 231-104-6 |
RTECSNumber | OM2100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.