Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium (Powder, 40-80 Mesh), Fisher Chemical

Catalog No. m7100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Color Silver
Melting Point 651.1°C
Atomic Weight 24.31
Quantity 100 g
Identification Pass Test
Mesh Size 40 to 80 Mesh
Packaging Glass Bottle
Form Solid
Formula Weight 24.31
Chemical Name or Material Magnesium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.