Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Magnesium Reagents for Hitachi™ Analyzers

Catalog No. p-318944 Shop All Pointe Scientific Products
Click to view available options
Quantity:
4 x 50mL R1, 4 x 50mL R2
5 x 250 R1mL
5 x 250 R2mL
Formulation:
DMSO (Dimethyl Sulfoxide) 20%, Xylidyl blue
R1: KCN 0.02% w/v, EGTA
Xylidyl blue 0.1mM, EGTA 0.13mM, DMSO 1.4M, Buffer, Surfactant, non-reactive Stabilizers including Potassium Cyanide at 0.02% w/v

Includes

Reagent 1 and Reagent 2 when combined: Xylidyl blue 0.1mM; EGTA 0.13mM; DMSO 1.4M; Buffer; surfactant; nonreactive stabilizers including potassium cyanide at 0.02% w/v

Requires

Calibrator

Quantify magnesium levels in serum

  • Method: Xylidyl Blue, Endpoint
  • Format: Liquid (ready to use)
  • Linearity: 4.86mg/dL
  • Expected Values: 1.6 to 3.0mg/dL

Applications:

For the quantitative determination of magnesium in serum with Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Magnesium Reagent Kit
Formulation Xylidyl blue 0.1mM, EGTA 0.13mM, DMSO 1.4M, Buffer, Surfactant, non-reactive Stabilizers including Potassium Cyanide at 0.02% w/v
For Use With (Application) Quantitative determination of Magnesium in Serum
Quantity 4 x 50mL R1, 4 x 50mL R2
Form Liquid

For in vitro diagnostic use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.