Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Ribbon, GSC Go Science Crazy
Shop All GSC Products


Click to view available options
Quantity:
25 g
Description
Magnesium ribbon 25 grams, is primarily used by lighting it on fire, which creates a bright, white light, intense heat, and magnesium oxide. Use this product to explore chemical reactions. For your safety, store in a cool, dry place, keep a fire extinguisher on hand, and do not look at the flame for too long.
Specifications
Specifications
Melting Point | 923 K (1202°F) |
Quantity | 25 g |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction